Learn More
Aviva Systems Biology Corp OLFML2B Antibody - N-terminal region (ARP47159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OLFML2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OLFML2B. Alternate names: MGC51337, RP11-227F8.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53508