Learn More
Aviva Systems Biology Corp Olfr329-ps Antibody - middle region (ARP59825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Olfr329-ps. It was validated on Western Blot. . Alternate names: MOR275-6P, Olfr329ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-845