Learn More
Cayman Chemical olIc ACDhydroxy oloyl Leu 1mg
SDP
Catalog No. 502895057
Encompass_Preferred
An internal standard for the quantification of oleic acid-hydroxy oleoyl leucine by GC- or LC-MS
Catalog No. 50-289-5057 Supplier Cayman Chemical Supplier No. 90043321 MG
An internal standard for the quantification of oleic acid-hydroxy oleoyl leucine by GC- or LC-MS