Learn More
IBL America OLIG2 Anti-Human Rabbit IgG Affinity Purify
Supplier:  IBL America 18953
Encompass_Preferred
For research use only, not for use in diagnostic procedures.This antibody can be stained in formalin fixed paraffin embedded tissues aftermicrowave pretreatment (10 min, 10mM Citrate Buffer, pH 6.0) by several Immunohistochemical techniques such as Avidin Bition Complex (ABC) method.The optimal dilution is about 0.1~1 μg/ml, however, the dilution rate should be optimized by each laboratories. This antibody can be used for western blotting in concentration of about 2μg/ml.
Catalog No. NC1347675