Learn More
Aviva Systems Biology Corp OLIG2 Antibody - N-terminal region (ARP31464_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OLIG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OLIG2. Alternate names: BHLHB1, OLIGO2, PRKCBP2, RACK17, bHLHe19ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-54235