Learn More
Aviva Systems Biology Corp OLIG3 Antibody - N-terminal region (ARP40015_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40015_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OLIG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OLIG3. Alternate names: Bhlhb7, bHLHe20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57264