Learn More
Agilent Technologies OLIGEL ssDNA Gel, Liquid, 250 mL
Catalog No. NC9962753
Encompass
OLIGEL ssDNA Gel is a pre-prepared liquid reagent designed for the separation and analysis of ssDNA and ssRNA using the Oligo Pro II system. This gel is formulated for direct, dye-free capillary gel electrophoresis to achieve high-resolution analysis of oligonucleotide purity and length. By containing a denaturant, it effectively unwinds secondary structures, ensuring consistent results independent of sequence or size, making it suitable for quality control of synthetic oligos and CRISPR sgRNA.
  • Achieves n-1 resolution for oligonucleotides up to 60 nt in length.
  • Pre-prepared format simplifies system setup and improves laboratory throughput.
  • Allows direct detection of ssDNA and ssRNA without the need for dye labeling.
  • Enables clear separation of 19, 20, 39, 40, 59, and 60 nt length peaks.
  • Reliable for purity assessment prior to critical applications.
Catalog No. NC9962753 Supplier Agilent Technologies Supplier No. DN4150250
OLIGEL ssDNA Gel is a pre-prepared liquid reagent designed for the separation and analysis of ssDNA and ssRNA using the Oligo Pro II system. This gel is formulated for direct, dye-free capillary gel electrophoresis to achieve high-resolution analysis of oligonucleotide purity and length. By containing a denaturant, it effectively unwinds secondary structures, ensuring consistent results independent of sequence or size, making it suitable for quality control of synthetic oligos and CRISPR sgRNA.
  • Achieves n-1 resolution for oligonucleotides up to 60 nt in length.
  • Pre-prepared format simplifies system setup and improves laboratory throughput.
  • Allows direct detection of ssDNA and ssRNA without the need for dye labeling.
  • Enables clear separation of 19, 20, 39, 40, 59, and 60 nt length peaks.
  • Reliable for purity assessment prior to critical applications.