Learn More
Zymo Research Corporation Oligo Binding Buffer, 40 ml
SDP
Catalog No. NC0915023
Encompass_Preferred
Oligo Binding Buffer (40 ml). For use in the Oligo Clean & Concentrator™ kits. Sold individually or as part of the Oligo Clean & Concentrator™ Kit.
Catalog No. NC0915023 Supplier Zymo Research Corporation Supplier No. D4060140
Oligo Binding Buffer (40 ml). For use in the Oligo Clean & Concentrator™ kits. Sold individually or as part of the Oligo Clean & Concentrator™ Kit.