Learn More
Research Products International Corp Oligo Clean & Concentrator, 200 Preps
SDP
Supplier:  Research Products International Corp ZD4061
Encompass
The Oligo Clean & Concentrator provides a streamlined method for efficient recovery and clean-up of DNA/RNA fragments and oligonucletides ≥ 16 nt from labeling (radioactive, biotin, DIG, etc.) and other enzymatic reactions. Unincorporated nucleotides, short oligos, dyes, enzymes, and salts are effectively removed by the clean-up procedure. DNA/RNA is washed and concentrated into a small volume of water. Purified DNA/RNA, available in just 2 minutes, is suitable for hybridization, gel shift assays, enzymatic reactions, ligation, sequencing, microarray analysis, etc.
Fast & Simple: Clean and concentrate DNA and RNA oligonucleotides in 2 minutes.
Ultra-Pure: Ready for hybridization, Next-Gen Sequencing, PCR, ligations, etc.
Complete removal: Eliminate dyes, salts, enzymes, nucleotides, and short oligos.
Catalog No. 50-197-7249