Learn More
Bioworld OLIGO CLEAN-UP AND CONCENTR
Catalog No. 50196941
Encompass_Preferred
OLIGO CLEAN-UP AND CONCENTRATION KIT: Fast and simple clean-up and concentration of oligonucleotides without the use of phenol Cleans and concentrates single-stranded or double-stranded DNA or RNA oligonucleotides larger than10bp; Rapid...
Catalog No. 50-196-941 Supplier Bioworld Supplier No. 107600841
OLIGO CLEAN-UP AND CONCENTRATION KIT: Fast and simple clean-up and concentration of oligonucleotides without the use of phenol Cleans and concentrates single-stranded or double-stranded DNA or RNA oligonucleotides larger than10bp; Rapid...