Learn More
New England Biolabs, Inc. Oligo d(T)18 mRNA Primer – 5 A260 units
SDP
Catalog No. 50812636
Encompass
Oligo d(T)18 mRNA primer is used for the priming and sequencing of mRNA adjacent to the 3'-poly A tail or tailed cDNA.
Catalog No. 50-812-636 Supplier New England Biolabs, Inc. Supplier No. S1316S
Oligo d(T)18 mRNA primer is used for the priming and sequencing of mRNA adjacent to the 3'-poly A tail or tailed cDNA.