Learn More
New England Biolabs, Inc. Oligo d(T)23VN – 1 A260 units
SDP
Supplier:  New England Biolabs, Inc. S1327S
Encompass
Oligo d(T)23 VN is used for the priming and sequencing of mRNA adjacent to the 3'-poly A tail. V = A or G or C N = A or G or C or T
Catalog No. 50-195-9366