Learn More
New England Biolabs, Inc. Oligo d(T)25 Magnetic Beads – 5 ml
SDP
Catalog No. 50995017
Encompass
Oligo d(T)25 is covalently coupled to 1 micron superparamagnetic particle through a linkage that is stable and leak resistant over a wide pH range, thereby permitting the small-scale isolation of mRNA from crude cell lysates and tissue. The isolation occurs through the hybridization of covalently coupled oligo d(T)25 to the poly(A) region present in most eukaryotic mRNAs.
Catalog No. 50-995-017 Supplier New England Biolabs, Inc. Supplier No. S1419S
Oligo d(T)25 is covalently coupled to 1 micron superparamagnetic particle through a linkage that is stable and leak resistant over a wide pH range, thereby permitting the small-scale isolation of mRNA from crude cell lysates and tissue. The isolation occurs through the hybridization of covalently coupled oligo d(T)25 to the poly(A) region present in most eukaryotic mRNAs.