Learn More
Medchemexpress LLC Oligodeoxynucleotide ODN 1585 (sodium salt) | 92.5% | 6430 (free acid) | DNA, d(G-sp-G-G-G-T-C-A-A-C-G-T-T-G-A-G-G-sp-G-sp-G-sp-G-sp-G) | 10 MG
SDP
Catalog No. 5000287150
Encompass_Preferred
ODN 1585 sodium is a synthetic CpG-containing oligodeoxynucleotide (sodium salt) used as an immunostimulatory reagent in research. It potently induces interferon and TNFα, enhances NK and CD8+ T-cell responses, and has applications in vaccine adjuvant studies and disease models such as acute myelogenous leukemia and malaria. Supplied as a white to off-white solid; sequence and analytical data are provided by the manufacturer.
Catalog No. 50-002-87150 Supplier Medchemexpress LLC Supplier No. HY150725C10MG
ODN 1585 sodium is a synthetic CpG-containing oligodeoxynucleotide (sodium salt) used as an immunostimulatory reagent in research. It potently induces interferon and TNFα, enhances NK and CD8+ T-cell responses, and has applications in vaccine adjuvant studies and disease models such as acute myelogenous leukemia and malaria. Supplied as a white to off-white solid; sequence and analytical data are provided by the manufacturer.