Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oligodeoxynucleotide ODN 2216 (CpG oligodeoxynucleotide) | 332437-00-0 | 97.9% | 6,432.00 Da | 10 MG
SDP

Catalog No. 5000286870
Encompass_Preferred

ODN 2216 is a Class A CpG oligodeoxynucleotide that acts as a human-specific Toll-like receptor 9 (TLR9) ligand and research reagent. It elicits robust type I interferon responses from plasmacytoid dendritic cells, promotes dendritic cell cytokine production, and can indirectly stimulate IFN-γ production in peripheral blood mononuclear cells.

  • Human-specific TLR9 agonist that induces type I interferons.
  • Stimulates IFN-α and IFN-β production by plasmacytoid dendritic cells.
  • Promotes IL-12 production by dendritic cells and indirect IFN-γ production in PBMCs.
  • Sequence: d(G-sp-G-sp-G-G-G-A-C-G-A-T-C-G-T-C-G-sp-G-sp-G-sp-G-sp-G-sp-G).
  • Molecular weight approximately 6,432 Da and purity ~97.9%.
  • Supplied as a 10 mg package; store sealed and away from moisture (in solvent: -80°C up to 6 months, -20°C up to 1 month).

Catalog No. 50-002-86870 Supplier Medchemexpress LLC Supplier No. HY15074110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ODN 2216 is a Class A CpG oligodeoxynucleotide that acts as a human-specific Toll-like receptor 9 (TLR9) ligand and research reagent. It elicits robust type I interferon responses from plasmacytoid dendritic cells, promotes dendritic cell cytokine production, and can indirectly stimulate IFN-γ production in peripheral blood mononuclear cells.

  • Human-specific TLR9 agonist that induces type I interferons.
  • Stimulates IFN-α and IFN-β production by plasmacytoid dendritic cells.
  • Promotes IL-12 production by dendritic cells and indirect IFN-γ production in PBMCs.
  • Sequence: d(G-sp-G-sp-G-G-G-A-C-G-A-T-C-G-T-C-G-sp-G-sp-G-sp-G-sp-G-sp-G).
  • Molecular weight approximately 6,432 Da and purity ~97.9%.
  • Supplied as a 10 mg package; store sealed and away from moisture (in solvent: -80°C up to 6 months, -20°C up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.