Learn More
QA Bio Inc Ludger Oligomannose-6 - 20µg
Supplier:  QA Bio Inc CNMAN620U
Encompass
Man-6 Glycan Synonyms: Man-6 N-linked oligosaccharide, Oligomannose 6 glycanDescription: Oligomannose N-linked oligosaccharide with 6 mannosyl residues. • Sources: Man-6 glycan is found in a wide range of both plant and animal glycoproteins. This product is typically purified from the oligosaccharide pool released from porcine thyroglobulin by hydrazinolysis using a combination of HPLC and glycosidase digestion. • Form: Dry. Dried by centrifugal evaporation from an aqueous solution. • Molecular Weight: 1398 • Purity: > 90% pure as assessed by a combination of 1 H-NMR and HPLC.
Catalog No. 50-223-6595