Learn More
Enzo Life Sciences Oligomycin (10mg). CAS: 1404-19-9
SDP
Catalog No. 502008922
Encompass_Preferred
Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1), preventing phosphoryl group transfer. Induces apoptosis. Purity: ≥90% (HPLC, TLC). Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol, methanol (10mg/ml) or acetone (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8922 Supplier Enzo Life Sciences Supplier No. ALX380037M010
Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1), preventing phosphoryl group transfer. Induces apoptosis. Purity: ≥90% (HPLC, TLC). Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol, methanol (10mg/ml) or acetone (50mg/ml). Long Term Storage: +4°C.