Learn More
Medchemexpress LLC Oligomycin | 1404-19-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN678210MG
Encompass_Preferred
Oligomycin is an antifungal antibiotic that acts as an inhibitor of H+-ATP-synthase. It blocks oxidative phosphorylation and the electron transport chain. Additionally, Oligomycin inhibits HIF-1alpha expression in hypoxic tumor cells. For research use only, it is not sold to patients. The product appears as a solid, white to off-white in color.
  • Antifungal antibiotic
  • Inhibitor of H+-ATP-synthase
  • Blocks oxidative phosphorylation
  • Blocks the electron transport chain
  • Inhibits HIF-1alpha expression in hypoxic tumor cells
  • Suppresses TNF-induced apoptosis by inhibiting the F0 part of H+-ATP-synthase
  • Sourced from microorganisms, specifically Streptomyces
  • Solid, white to off-white appearance
Catalog No. 50-004-19338