Learn More
Medchemexpress LLC HY-N6782 5mg , Oligomycin CAS:1404-19-9 Purity:>98%
SDP
Catalog No. 502259407
Encompass_Preferred
HY-N6782 5mg Oligomycin CAS: 1404-19-9 Oligomycin, an antifungal antibiotic, is an inhibitor of H+-ATP-synthase. Oligomycin blocks oxidative phosphorylation and the electron transport chain. Oligomycin inhibits HIF-1alpha expression in hypoxic tumor cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9407 Supplier Medchemexpress LLC Supplier No. HYN67825MG
HY-N6782 5mg Oligomycin CAS: 1404-19-9 Oligomycin, an antifungal antibiotic, is an inhibitor of H+-ATP-synthase. Oligomycin blocks oxidative phosphorylation and the electron transport chain. Oligomycin inhibits HIF-1alpha expression in hypoxic tumor cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.