Learn More
Enzo Life Sciences Oligomycin A (25mg). CAS: 579-13-5
SDP
Catalog No. 502010415
Encompass_Preferred
Inhibits mitochondrial ATPases (F1F0). Stimulates lysosome acidification due to shunting of membrane potential. Induces apoptosis in a variety of cell types. Induces autophagy in the IPLB-LdFB insect cell line. Purity: ≥97% (HPLC, TLC). Solubility: Soluble in DMSO (300mg/ml), 100% ethanol (200mg/ml) or acetone (800mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0415 Supplier Enzo Life Sciences Supplier No. BMLCM1110025
Inhibits mitochondrial ATPases (F1F0). Stimulates lysosome acidification due to shunting of membrane potential. Induces apoptosis in a variety of cell types. Induces autophagy in the IPLB-LdFB insect cell line. Purity: ≥97% (HPLC, TLC). Solubility: Soluble in DMSO (300mg/ml), 100% ethanol (200mg/ml) or acetone (800mg/ml). Long Term Storage: -20°C.