Learn More
Medchemexpress LLC Oligomycin A | 579-13-5 | 99.9% | 791.06 | 1 MG
SDP
Catalog No. 5000394494
Encompass_Preferred
Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor with a Ki of 1 μM. It also demonstrates anti-fungal activity and is for research use only.
  • Mitochondrial F0F1-ATPase inhibitor (Ki: 1 μM)
  • Shows anti-fungal activity
  • Inhibits the activity of Triton X-100-solubilized ATPase (Ki of 0.1 μM)
  • Cytotoxic to NCI-60 cell lines (GI50 of 10 nM)
  • Inhibits the ability of spermatozoa to achieve feasible in vitro capacitation (IVC) at 2.4 μM)
  • Suppresses progesterone-induced in vitro acrosome exocytosis (IVAE) and concomitant peaks of O2 consumption and ATP levels
Catalog No. 50-003-94494 Supplier Medchemexpress LLC Supplier No. HY165891MG
Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor with a Ki of 1 μM. It also demonstrates anti-fungal activity and is for research use only.
  • Mitochondrial F0F1-ATPase inhibitor (Ki: 1 μM)
  • Shows anti-fungal activity
  • Inhibits the activity of Triton X-100-solubilized ATPase (Ki of 0.1 μM)
  • Cytotoxic to NCI-60 cell lines (GI50 of 10 nM)
  • Inhibits the ability of spermatozoa to achieve feasible in vitro capacitation (IVC) at 2.4 μM)
  • Suppresses progesterone-induced in vitro acrosome exocytosis (IVAE) and concomitant peaks of O2 consumption and ATP levels