Learn More
Medchemexpress LLC Oligomycin A | 579-13-5 | 99.9% | 791.06 | 1 ML
SDP
Catalog No. 5000394515
Encompass_Preferred
Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor with a Ki of 1 μM. It also shows anti-fungal activity, is cytotoxic to NCI-60 cell lines (GI50 of 10 nM), and inhibits the activity of Triton X-100-solubilized ATPase. It suppresses progesterone-induced in vitro acrosome exocytosis (IVAE) and concomitant O2 consumption and ATP levels.
  • Acts as a mitochondrial F0F1-ATPase inhibitor
  • Shows anti-fungal activity
  • Cytotoxic to NCI-60 cell lines
  • Inhibits activity of Triton X-100-solubilized ATPase
  • Suppresses progesterone-induced in vitro acrosome exocytosis (IVAE)
  • Inhibits spermatozoa's ability to achieve feasible in vitro capacitation (IVC)
Catalog No. 50-003-94515 Supplier Medchemexpress LLC Supplier No. HY1658910MM/1ML
Add to Cart
Add to Cart
Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor with a Ki of 1 μM. It also shows anti-fungal activity, is cytotoxic to NCI-60 cell lines (GI50 of 10 nM), and inhibits the activity of Triton X-100-solubilized ATPase. It suppresses progesterone-induced in vitro acrosome exocytosis (IVAE) and concomitant O2 consumption and ATP levels.
  • Acts as a mitochondrial F0F1-ATPase inhibitor
  • Shows anti-fungal activity
  • Cytotoxic to NCI-60 cell lines
  • Inhibits activity of Triton X-100-solubilized ATPase
  • Suppresses progesterone-induced in vitro acrosome exocytosis (IVAE)
  • Inhibits spermatozoa's ability to achieve feasible in vitro capacitation (IVC)