Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oligomycin A | 579-13-5 | MFCD00065705 | 99.9% | 791.06 | C45H74O11 | 10 MG
SDP

Catalog No. 5000288919
Encompass_Preferred

Oligomycin A is a Streptomyces-derived macrolide antibiotic that potently inhibits mitochondrial ATP synthase (F0F1-ATPase). Supplied as a high-purity powder for research use, it is used to block oxidative phosphorylation in biochemical and cellular assays, probe mitochondrial function, and support in vivo formulations when properly solubilized.

  • High purity ≥99.9% (HPLC).
  • Molecular formula C45H74O11; molecular weight 791.06.
  • White to off-white powder; sparingly soluble in water, soluble in DMSO and ethanol.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Applications include mitochondrial respiration assays, apoptosis studies, and antifungal research.

Catalog No. 50-002-88919 Supplier Medchemexpress LLC Supplier No. HY1658910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oligomycin A is a Streptomyces-derived macrolide antibiotic that potently inhibits mitochondrial ATP synthase (F0F1-ATPase). Supplied as a high-purity powder for research use, it is used to block oxidative phosphorylation in biochemical and cellular assays, probe mitochondrial function, and support in vivo formulations when properly solubilized.

  • High purity ≥99.9% (HPLC).
  • Molecular formula C45H74O11; molecular weight 791.06.
  • White to off-white powder; sparingly soluble in water, soluble in DMSO and ethanol.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
  • Applications include mitochondrial respiration assays, apoptosis studies, and antifungal research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.