Learn More
TARGETMOL CHEMICALS INC OLIGOMYCIN A 5MG
SDP
Catalog No. 502576531
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oligomycin A (MCH 32) is an inhibitor of ATP synthase inhibits oxidative phosphorylation and all the ATP-dependent processes occurring on the coupling membrane of mitochondria. purity: 99%
Catalog No. 50-257-6531 Supplier TARGETMOL CHEMICALS INC Supplier No. T63235MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oligomycin A (MCH 32) is an inhibitor of ATP synthase inhibits oxidative phosphorylation and all the ATP-dependent processes occurring on the coupling membrane of mitochondria. purity: 99%