Learn More
Cayman Chemical OlIgomycIn B 10mg
SDP
Catalog No. 502954646
Encompass_Preferred
A nonselective inhibitor of the mitochondrial F1FO ATP synthase that can reduce the rate of ATP depletion in myocardial ischemia and decrease calcium-induced calcium release oscillation frequency of rat sensory neurons
Catalog No. 50-295-4646 Supplier Cayman Chemical Supplier No. 1134310 MG
A nonselective inhibitor of the mitochondrial F1FO ATP synthase that can reduce the rate of ATP depletion in myocardial ischemia and decrease calcium-induced calcium release oscillation frequency of rat sensory neurons