Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oligomycin B | 11050-94-5 | 1 MG
SDP

Catalog No. 5000417924
Encompass_Preferred

Oligomycin B is an antibiotic isolated from marine Streptomyces. It functions as an eukaryotic ATP synthase inhibitor and induces apoptosis.

  • Purity of 97.67%
  • Molecular weight of 805.05
  • Molecular formula C₄₅H₇₂O₁₂
  • Appearance as a solid
  • Color is white to off-white
  • Classified as an antibiotic, for disease research, antifungal, and macrolide antibiotic
  • Isolated from marine Streptomyces microorganisms
  • Shipping at room temperature (continental US)
  • Store at -20°C, protected from light and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Catalog No. 50-004-17924 Supplier Medchemexpress LLC Supplier No. HYN67841MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oligomycin B is an antibiotic isolated from marine Streptomyces. It functions as an eukaryotic ATP synthase inhibitor and induces apoptosis.

  • Purity of 97.67%
  • Molecular weight of 805.05
  • Molecular formula C₄₅H₇₂O₁₂
  • Appearance as a solid
  • Color is white to off-white
  • Classified as an antibiotic, for disease research, antifungal, and macrolide antibiotic
  • Isolated from marine Streptomyces microorganisms
  • Shipping at room temperature (continental US)
  • Store at -20°C, protected from light and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.