Learn More
Lkt Laboratories Oligomycin B, ≥97%, 1MG
SDP
Supplier:  Lkt Laboratories O45321MG
Encompass_Preferred
Macrolide; F1F0 ATP synthase inhibitor. CAS: 11050-94-5. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4979