Learn More
Enzo Life Sciences Oligomycin C (1mg). CAS: 11052-72-5
SDP
Supplier:  Enzo Life Sciences ALX380039M001
Encompass_Preferred
Minor component of the oligomycin complex. Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1F0), preventing phosphoryl group transfer. Induces apoptosis. Alternative name: 12-Deoxyoligomycin A. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8924