Learn More
Cayman Chemical OlIgomycIn D 2.5mg
SDP
Catalog No. 502985295
Encompass_Preferred
A macrolide antibiotic that inhibits the mitochondrial F1FOATPase; inhibits K-Ras plasma membrane localization in MDCK cells with an IC50 value of 3.49 nM and is cyctoxic to SW620 colon cancer cells with an IC50 value of 36 µM
Catalog No. 50-298-5295 Supplier Cayman Chemical Supplier No. 201842.5 MG
A macrolide antibiotic that inhibits the mitochondrial F1FOATPase; inhibits K-Ras plasma membrane localization in MDCK cells with an IC50 value of 3.49 nM and is cyctoxic to SW620 colon cancer cells with an IC50 value of 36 µM