Learn More
Abcam Oligonucleotide Conjugation Kit, 3 x 100UG
Catalog No. 502587888
Encompass_Preferred
Abcam's Oligonucleotide Conjugation Kit (ab218260) offers several standout features: - Rapid Conjugation: enables generation of antibody-oligonucleotide conjugates in less than 2 hours. - High Efficiency: achieves high levels of antibody and oligonucleotide recovery, with conjugation efficiency ~ 95-100%. - Versatility: Suitable for conjugating antibodies to single-stranded oligonucleotides (10-120 bases) and double-stranded oligonucleotides (up to 80 bases), providing flexibility for various research needs
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-7888 Supplier Abcam Supplier No. AB2182603X100UG
Abcam's Oligonucleotide Conjugation Kit (ab218260) offers several standout features: - Rapid Conjugation: enables generation of antibody-oligonucleotide conjugates in less than 2 hours. - High Efficiency: achieves high levels of antibody and oligonucleotide recovery, with conjugation efficiency ~ 95-100%. - Versatility: Suitable for conjugating antibodies to single-stranded oligonucleotides (10-120 bases) and double-stranded oligonucleotides (up to 80 bases), providing flexibility for various research needs
The product is subject to the following: Abcam Restricted Use Statement