Learn More
Medchemexpress LLC Oligopeptide-24 (CG-EDP3) | 1018685-16-9 | 99.6% | 1271.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP52575MG
Encompass_Preferred
Oligopeptide-24 (CG-EDP3) is a 13 amino acid biomimetic peptide designed for skin repair. It upregulates the expression of elastin and hyaluronic acid while increasing fibroblast activity. This peptide can be utilized in cosmetic formulations as an anti-wrinkle and firming agent, and it has the potential to boost endogenous growth factor concentration in the skin significantly.
  • Upregulates elastin and hyaluronic acid expression
  • Increases fibroblast activity
  • Functions as an anti-wrinkle and firming agent
  • Increases endogenous growth factor concentration in skin
  • Can be accurately quantified in cosmetic products
Catalog No. 50-003-98929