Learn More
Medchemexpress LLC Oligopeptide P11-4 | 593266-60-5 | 98.5% | 1595.67 | 10 MG
SDP
Catalog No. 5000425446
Encompass_Preferred
Oligopeptide P11-4 is an α-peptide capable of self-assembling into β-sheet amyloids, exhibiting a hydrogel appearance at low pH. This peptide is utilized in biomimetic mineralization, enamel regeneration, and as an oral care agent.
  • Self-assembles into β-sheet amyloids
  • Exhibits hydrogel appearance at low pH
  • Used in biomimetic mineralization
  • Used for enamel regeneration
  • Functions as an oral care agent
Catalog No. 50-004-25446 Supplier Medchemexpress LLC Supplier No. HYP1029410MG
Oligopeptide P11-4 is an α-peptide capable of self-assembling into β-sheet amyloids, exhibiting a hydrogel appearance at low pH. This peptide is utilized in biomimetic mineralization, enamel regeneration, and as an oral care agent.
  • Self-assembles into β-sheet amyloids
  • Exhibits hydrogel appearance at low pH
  • Used in biomimetic mineralization
  • Used for enamel regeneration
  • Functions as an oral care agent