Learn More
Medchemexpress LLC Oligopeptide P11-4 | 593266-60-5 | 98.5% | 1595.67 | 25 MG
SDP
Catalog No. 5000426453
Encompass_Preferred
Oligopeptide P11-4 is an α-peptide that can self-assemble into β-sheet amyloids with a hydrogel appearance at low pH. It can be used in biomimetic mineralization, enamel regeneration, and as an oral care agent.
  • Self-assembles into β-sheet amyloids with a hydrogel appearance at low pH
  • Used in biomimetic mineralization
  • Used in enamel regeneration
  • Used as an oral care agent
Catalog No. 50-004-26453 Supplier Medchemexpress LLC Supplier No. 25MGHYP10294
Oligopeptide P11-4 is an α-peptide that can self-assemble into β-sheet amyloids with a hydrogel appearance at low pH. It can be used in biomimetic mineralization, enamel regeneration, and as an oral care agent.
  • Self-assembles into β-sheet amyloids with a hydrogel appearance at low pH
  • Used in biomimetic mineralization
  • Used in enamel regeneration
  • Used as an oral care agent