Learn More
Medchemexpress LLC Oligopeptide P11-4 | 593266-60-5 | 98.5% | 1595.67 | 5 MG
SDP
Catalog No. 5000424938
Encompass_Preferred
Oligopeptide P11-4 is an α-peptide that can self-assemble into β-sheet amyloids with a hydrogel appearance at low pH. It is utilized in various applications.
  • Used in biomimetic mineralization.
  • Used in enamel regeneration.
  • Used as an oral care agent.
Catalog No. 50-004-24938 Supplier Medchemexpress LLC Supplier No. HYP10294100MG
Oligopeptide P11-4 is an α-peptide that can self-assemble into β-sheet amyloids with a hydrogel appearance at low pH. It is utilized in various applications.
  • Used in biomimetic mineralization.
  • Used in enamel regeneration.
  • Used as an oral care agent.