Learn More
Waters Corp SPE Cartridge, 1 cc, 60 mg Sorbent, 30 μm, WAX
Catalog No. 502886218 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The SPE 1 cc Cartridge is designed for oligonucleotide bioanalysis and contains 60 mg of OligoWorks SPE WAX Sorbent per cartridge. The sorbent is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, which is oligonucleotide tested and batch-selected for optimal oligonucleotide recovery.
  • Solid phase extraction product type
  • Oligonucleotide application
  • 50 units per package
  • WAX chemistry
  • Not endcapped
  • Classic cartridge format
  • Mass spec compatible
  • Mixed mode
  • 30 μm particle size
  • 80 Å pore size
  • Copolymer sorbent substrate
  • 60 mg sorbent weight
  • Water wettable
  • pH range 0 to 14
  • Polypropylene material
Catalog No. 50-288-6218 Supplier Waters Corp Supplier No. 186010751
The SPE 1 cc Cartridge is designed for oligonucleotide bioanalysis and contains 60 mg of OligoWorks SPE WAX Sorbent per cartridge. The sorbent is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, which is oligonucleotide tested and batch-selected for optimal oligonucleotide recovery.
  • Solid phase extraction product type
  • Oligonucleotide application
  • 50 units per package
  • WAX chemistry
  • Not endcapped
  • Classic cartridge format
  • Mass spec compatible
  • Mixed mode
  • 30 μm particle size
  • 80 Å pore size
  • Copolymer sorbent substrate
  • 60 mg sorbent weight
  • Water wettable
  • pH range 0 to 14
  • Polypropylene material