Learn More
Waters Corp OligoWorks SPE Cartridge, 3 cc, 60 mg Sorbent, 30 μm
Catalog No. 502886197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The OligoWorks SPE 3 cc Cartridge is engineered for oligonucleotide bioanalysis applications. Each cartridge contains 60 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. This sorbent undergoes rigorous oligonucleotide testing and batch-selection to ensure optimal recovery of oligonucleotides.
  • Utilizes WAX chemistry for solid phase extraction.
  • Features a classic cartridge format with 30 μm particle size.
  • Mass spectrometry compatible and operates in mixed mode.
  • Sorbent substrate is copolymer with 80 Å pore size.
  • Water wettable and constructed from polypropylene.
  • Stable at room temperature with a 2-year expiration.
  • Automation friendly for streamlined processes.
  • Supports a broad pH range from 0 to 14.
  • Recommended volumes for SPE protocol: 3 mL condition, 5 mL load, 4 mL wash, 4 mL elute.
Catalog No. 50-288-6197 Supplier Waters Corp Supplier No. 186010620
The OligoWorks SPE 3 cc Cartridge is engineered for oligonucleotide bioanalysis applications. Each cartridge contains 60 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. This sorbent undergoes rigorous oligonucleotide testing and batch-selection to ensure optimal recovery of oligonucleotides.
  • Utilizes WAX chemistry for solid phase extraction.
  • Features a classic cartridge format with 30 μm particle size.
  • Mass spectrometry compatible and operates in mixed mode.
  • Sorbent substrate is copolymer with 80 Å pore size.
  • Water wettable and constructed from polypropylene.
  • Stable at room temperature with a 2-year expiration.
  • Automation friendly for streamlined processes.
  • Supports a broad pH range from 0 to 14.
  • Recommended volumes for SPE protocol: 3 mL condition, 5 mL load, 4 mL wash, 4 mL elute.