Learn More
Waters Corp SPE Cartridge, 6 cc, 150 mg Sorbent, 30 μm Particle Size
Catalog No. 502886234 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge is specifically designed for oligonucleotide bioanalysis. It features 150 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that is oligonucleotide tested and batch-selected for optimal recovery.
  • Designed specifically for oligonucleotide bioanalysis.
  • Contains 150 mg of polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Sorbent is tested and batch-selected for optimal oligonucleotide recovery.
  • Simplifies sample preparation and improves robustness for greater MS sensitivity.
  • Offers water wettability, pH stability, and eliminates silanol interactions.
  • No conditioning or equilibration required for the sorbent prior to sample loading.
  • Produces clean, concentrated eluates for direct LC-MS analysis.
Catalog No. 50-288-6234 Supplier Waters Corp Supplier No. 186010753
This solid phase extraction cartridge is specifically designed for oligonucleotide bioanalysis. It features 150 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that is oligonucleotide tested and batch-selected for optimal recovery.
  • Designed specifically for oligonucleotide bioanalysis.
  • Contains 150 mg of polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Sorbent is tested and batch-selected for optimal oligonucleotide recovery.
  • Simplifies sample preparation and improves robustness for greater MS sensitivity.
  • Offers water wettability, pH stability, and eliminates silanol interactions.
  • No conditioning or equilibration required for the sorbent prior to sample loading.
  • Produces clean, concentrated eluates for direct LC-MS analysis.