Learn More
Waters Corp SPE Macroplate, 96-well, 60 mg Sorbent/well, OligoWorks WAX Sorbent
Catalog No. 502886184 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The OligoWorks SPE Macroplate is a 96-deepwell filter plate with 60 mg/well of OligoWorks WAX Sorbent. This polymeric mixed-mode sorbent performs solid phase extraction of oligonucleotides from biological matrices, providing enhanced cleanup, high recovery, and repeatability.
  • Dual retention from water-wettable HLB copolymer with piperazine WAX ligand.
  • No conditioning or equilibration required before sample loading.
  • Standardized and automation-friendly for oligonucleotide bioanalysis.
  • High recovery for diverse oligonucleotide therapeutic modalities.
  • Streamlined sample pretreatment reduces method development.
  • Clean, concentrated eluates for direct LC-MS analysis.
  • Room temperature stable with a 2-year expiration date.
Catalog No. 50-288-6184 Supplier Waters Corp Supplier No. 186010749
The OligoWorks SPE Macroplate is a 96-deepwell filter plate with 60 mg/well of OligoWorks WAX Sorbent. This polymeric mixed-mode sorbent performs solid phase extraction of oligonucleotides from biological matrices, providing enhanced cleanup, high recovery, and repeatability.
  • Dual retention from water-wettable HLB copolymer with piperazine WAX ligand.
  • No conditioning or equilibration required before sample loading.
  • Standardized and automation-friendly for oligonucleotide bioanalysis.
  • High recovery for diverse oligonucleotide therapeutic modalities.
  • Streamlined sample pretreatment reduces method development.
  • Clean, concentrated eluates for direct LC-MS analysis.
  • Room temperature stable with a 2-year expiration date.