Learn More
Waters Corp SPE Macroplate, 96-well, 30 mg Sorbent, Polypropylene
Catalog No. 502886209 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The SPE Macroplate is a 96-deepwell filter plate designed for solid phase extraction. Each well contains 30 mg of WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It is specifically batch-selected for optimal oligonucleotide selectivity and recovery, making it suitable for the extraction of oligonucleotides and related DNA/RNA therapeutics from complex biological matrices.
  • 96-deepwell plate with a 2 mL well volume.
  • 30 mg of WAX sorbent per well.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for oligonucleotide recovery and selectivity.
  • Water wettable.
  • Mass Spec compatible.
  • Constructed from polypropylene.
  • Wide pH range from 0 to 14.
  • Does not require pre-conditioning or equilibration prior to sample loading.
  • Dual modes of retention based on anion-exchange interaction.
Catalog No. 50-288-6209 Supplier Waters Corp Supplier No. 186010619
The SPE Macroplate is a 96-deepwell filter plate designed for solid phase extraction. Each well contains 30 mg of WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It is specifically batch-selected for optimal oligonucleotide selectivity and recovery, making it suitable for the extraction of oligonucleotides and related DNA/RNA therapeutics from complex biological matrices.
  • 96-deepwell plate with a 2 mL well volume.
  • 30 mg of WAX sorbent per well.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for oligonucleotide recovery and selectivity.
  • Water wettable.
  • Mass Spec compatible.
  • Constructed from polypropylene.
  • Wide pH range from 0 to 14.
  • Does not require pre-conditioning or equilibration prior to sample loading.
  • Dual modes of retention based on anion-exchange interaction.