Learn More
Waters Corp OligoWorks SPE Macroplate, 96-Well, 10 mg Sorbent, Polypropylene
Catalog No. 502886247 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OligoWorks SPE Macroplate is a 96-deepwell filter plate with a 2 mL well volume. Each well contains 10 mg of OligoWorks SPE WAX Sorbent, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It is oligonucleotide tested and batch-selected for optimal oligonucleotide recovery.
  • 96-deepwell filter plate
  • 2 mL well volume
  • 10 mg sorbent per well
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Oligonucleotide tested
  • Batch-selected for optimal oligonucleotide recovery
  • Constructed from polypropylene
  • Mass spectrometry compatible
  • Water wettable
  • pH range of 0 to 14
Catalog No. 50-288-6247 Supplier Waters Corp Supplier No. 186010752
This OligoWorks SPE Macroplate is a 96-deepwell filter plate with a 2 mL well volume. Each well contains 10 mg of OligoWorks SPE WAX Sorbent, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It is oligonucleotide tested and batch-selected for optimal oligonucleotide recovery.
  • 96-deepwell filter plate
  • 2 mL well volume
  • 10 mg sorbent per well
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Oligonucleotide tested
  • Batch-selected for optimal oligonucleotide recovery
  • Constructed from polypropylene
  • Mass spectrometry compatible
  • Water wettable
  • pH range of 0 to 14