Learn More
Waters Corp OligoWorks SPE Microplate, 96-well, 2 mg Sorbent, WAX Sorbent
Catalog No. 502886194 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The OligoWorks SPE Microplate is a 96-well filter plate of standard height with a 400 mL well volume. Each well contains 2 mg of OligoWorks SPE WAX Sorbent. The sorbent is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been oligonucleotide tested and batch-selected.
  • Standard height and 400 mL well volume
  • Oligonucleotide tested
  • Batch-selected for optimal oligonucleotide recovery
  • 96 wells
  • 2 mg sorbent per well
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • WAX chemistry
  • Not endcapped
  • Mass spec compatible
  • Mixed mode operation
  • 30 μm particle size
  • 80 Å pore size
  • Copolymer sorbent substrate
  • Water wettable
  • pH range: 0 to 14 pH
  • Made of polypropylene
Catalog No. 50-288-6194 Supplier Waters Corp Supplier No. 186010618
The OligoWorks SPE Microplate is a 96-well filter plate of standard height with a 400 mL well volume. Each well contains 2 mg of OligoWorks SPE WAX Sorbent. The sorbent is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been oligonucleotide tested and batch-selected.
  • Standard height and 400 mL well volume
  • Oligonucleotide tested
  • Batch-selected for optimal oligonucleotide recovery
  • 96 wells
  • 2 mg sorbent per well
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • WAX chemistry
  • Not endcapped
  • Mass spec compatible
  • Mixed mode operation
  • 30 μm particle size
  • 80 Å pore size
  • Copolymer sorbent substrate
  • Water wettable
  • pH range: 0 to 14 pH
  • Made of polypropylene