Learn More
Medchemexpress LLC Olitigaltin | 1450824-22-2 | 99.7% | 648.64 | 1 ML
SDP
Catalog No. 5000401661
Encompass_Preferred
Olitigaltin (TD139) is a synthetic galectin-3 inhibitor with a high affinity for galectin-3, galectin-1, and galectin-7. It is intended for research use only. This product is provided as a 10 mM solution in DMSO, ready for reconstitution or direct use.
  • High affinity galectin-3 inhibitor
  • Targets galectin-1 and galectin-7
  • Provided as a 10 mM solution
  • Suitable for research use
Catalog No. 50-004-01661 Supplier Medchemexpress LLC Supplier No. HY1994010MM/1ML
Olitigaltin (TD139) is a synthetic galectin-3 inhibitor with a high affinity for galectin-3, galectin-1, and galectin-7. It is intended for research use only. This product is provided as a 10 mM solution in DMSO, ready for reconstitution or direct use.
  • High affinity galectin-3 inhibitor
  • Targets galectin-1 and galectin-7
  • Provided as a 10 mM solution
  • Suitable for research use