Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Olive Oil
SCI_ED

Catalog No. S25452 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25452 500 mL Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25452
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 8001-25-0
Synonym Olive oil, edible

Specifications

Density 0. 9135g/cmsuperscript_3;
Packaging Solution
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Olive Oil
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.