Learn More
MSC Oliver Footwear Boot, 150 mm, Water Resistant Leather, Lace Up, Steel Toe Cap
Catalog No. 5000509290
Encompass
This 150 mm black lace-up work boot from the AT 55 Series features a lightweight sole with a self-cleaning tread profile. It includes a plush satin feel lining and a built-in comfort system for enhanced wear.
  • Lightweight, self-cleaning sole.
  • Plush satin lining for moisture and odor control.
  • Integrated comfort system with shock absorbing polyurethane midsole.
  • Water resistant leather upper, lace locking fixture.
  • Kevlar reinforced seams for abrasion and chemical resistance.
  • Impact absorption system.
  • Toe bumper for scuff protection.
  • Durable, slip-resistant (SRC), heat-resistant (to 300°C) rubber outsole.
  • Sole resistant to oils and acids.
  • Anti-static.
  • Type 1 steel toe cap, 200 Joule tested.
  • Airport friendly with antimicrobial technology.
Catalog No. 50-005-09290 Supplier MSC Supplier No. 39053137
This 150 mm black lace-up work boot from the AT 55 Series features a lightweight sole with a self-cleaning tread profile. It includes a plush satin feel lining and a built-in comfort system for enhanced wear.
  • Lightweight, self-cleaning sole.
  • Plush satin lining for moisture and odor control.
  • Integrated comfort system with shock absorbing polyurethane midsole.
  • Water resistant leather upper, lace locking fixture.
  • Kevlar reinforced seams for abrasion and chemical resistance.
  • Impact absorption system.
  • Toe bumper for scuff protection.
  • Durable, slip-resistant (SRC), heat-resistant (to 300°C) rubber outsole.
  • Sole resistant to oils and acids.
  • Anti-static.
  • Type 1 steel toe cap, 200 Joule tested.
  • Airport friendly with antimicrobial technology.