Learn More
MSC Oliver Slip-On Work Shoe, Women's, Steel Toe, Water Resistant, Brown
Catalog No. 5000509351
Encompass
The 49 Series Slip-On Work Shoe is designed for women, offering robust toe protection and cushioning. These leather shoes meet or exceed ASTM F2413-11 F I/75 C/75 SD safety standards, featuring steel safety toe caps. The polyurethane rubber outsoles provide excellent heat, chemical, and slip resistance, plus electrical hazard protection.
  • Women's specific design for optimal fit.
  • High heat resistant and highly slip-resistant rubber outsole.
  • Cushiony PU midsole for energy return.
  • Wide steel toe cap for comfortable protection.
  • Anti-static performance.
  • Meets ASTM F2413-11 F I/75 C/75 SD safety standard.
  • Resistant to chemicals, oils, and abrasions.
  • Electrical hazard rated.
  • Water resistant leather uppers.
Catalog No. 50-005-09351 Supplier MSC Supplier No. 39051685
The 49 Series Slip-On Work Shoe is designed for women, offering robust toe protection and cushioning. These leather shoes meet or exceed ASTM F2413-11 F I/75 C/75 SD safety standards, featuring steel safety toe caps. The polyurethane rubber outsoles provide excellent heat, chemical, and slip resistance, plus electrical hazard protection.
  • Women's specific design for optimal fit.
  • High heat resistant and highly slip-resistant rubber outsole.
  • Cushiony PU midsole for energy return.
  • Wide steel toe cap for comfortable protection.
  • Anti-static performance.
  • Meets ASTM F2413-11 F I/75 C/75 SD safety standard.
  • Resistant to chemicals, oils, and abrasions.
  • Electrical hazard rated.
  • Water resistant leather uppers.