Learn More
Cayman Chemical OlIvetolIc AcId 100mg
SDP
Catalog No. 502934090
Encompass_Preferred
An analytical reference standard categorized as an intermediate in the phytocannabinoid biosynthetic pathway; a precursor in the synthesis of CBGA, Δ9-THC, and CBD; intended for research and forensic applications
Catalog No. 50-293-4090 Supplier Cayman Chemical Supplier No. 26282100 MG
An analytical reference standard categorized as an intermediate in the phytocannabinoid biosynthetic pathway; a precursor in the synthesis of CBGA, Δ9-THC, and CBD; intended for research and forensic applications