Learn More
Medchemexpress LLC Olivetolic acid | 491-72-5 | 99.9% | 224.25 g/mol | C12H16O4 | 25 MG
SDP
Catalog No. 5000208902
Encompass_Preferred
Olivetolic acid is a biosynthetic precursor of cannabinol acid (CBGA) used as a research reagent for studies of cannabinoid biosynthesis and anticonvulsant effects. It is supplied as a white to off-white solid with high purity for analytical and in vitro applications.
  • Provides a biosynthetic precursor to cannabinoid acid (CBGA).
  • Supports research on anticonvulsant activity and convulsion models.
  • High purity suitable for analytical and biochemical assays.
  • Available in small pack sizes such as 25 mg for lab use.
  • Solid, white to off-white appearance for easy handling and storage.
  • Recommended storage: 4°C sealed, avoid moisture; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-08902 Supplier Medchemexpress LLC Supplier No. 25MGHYW090292
Add to Cart
Add to Cart
Olivetolic acid is a biosynthetic precursor of cannabinol acid (CBGA) used as a research reagent for studies of cannabinoid biosynthesis and anticonvulsant effects. It is supplied as a white to off-white solid with high purity for analytical and in vitro applications.
  • Provides a biosynthetic precursor to cannabinoid acid (CBGA).
  • Supports research on anticonvulsant activity and convulsion models.
  • High purity suitable for analytical and biochemical assays.
  • Available in small pack sizes such as 25 mg for lab use.
  • Solid, white to off-white appearance for easy handling and storage.
  • Recommended storage: 4°C sealed, avoid moisture; in solvent -80°C (6 months) or -20°C (1 month).