Learn More
Cayman Chemical Olivetolic Acid, Purity: ≥90% (mixture of isomers), CAS: 491-72-5, 50 mg
SDP
Catalog No. 502935107
Encompass_Preferred
An analytical reference standard categorized as an intermediate in the phytocannabinoid biosynthetic pathway; a precursor in the synthesis of CBGA, Δ9-THC, and CBD; intended for research and forensic applications
Catalog No. 50-293-5107 Supplier Cayman Chemical Supplier No. 2628250 MG
An analytical reference standard categorized as an intermediate in the phytocannabinoid biosynthetic pathway; a precursor in the synthesis of CBGA, Δ9-THC, and CBD; intended for research and forensic applications