Learn More
Medchemexpress LLC HY-17004 100mg Medchemexpress, Olmesartan CAS:144689-24-7 Purity:>98%
SDP
Catalog No. 502081925
Encompass_Preferred
Medchemexpress, HY-17004 100mg Olmesartan CAS:144689-24-7 Olmesartan (RNH-6270) is an angiotensin II receptor (AT1R) antagonist used to treat high blood pressure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1925 Supplier Medchemexpress LLC Supplier No. HY17004100MG
Medchemexpress, HY-17004 100mg Olmesartan CAS:144689-24-7 Olmesartan (RNH-6270) is an angiotensin II receptor (AT1R) antagonist used to treat high blood pressure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.