Learn More
Medchemexpress LLC Olmesartan | 144689-24-7 | 99.9% | 446.50 | 50 MG
SDP
Catalog No. 5000390507
Encompass_Preferred
Olmesartan, also known as RNH-6270, is an angiotensin II receptor (AT1R) antagonist that is utilized in the treatment of high blood pressure. It is intended for research use only.
  • It is an angiotensin II receptor (AT1R) antagonist.
  • It is used to treat high blood pressure.
  • It inhibits the growth of HeLa cells in a concentration- and time-dependent manner.
  • Repeated dosing decreases mean arterial blood pressure without significantly influencing body weight or food intake.
  • Treatment inhibits cardiac hypertrophy.
  • Treatment reverses decreased gene expressions of ACE2 and Mas receptor, and inhibits enhanced NADPH oxidase (Nox)4 expression and reactive oxygen species.
Catalog No. 50-003-90507 Supplier Medchemexpress LLC Supplier No. HY1700450MG
Olmesartan, also known as RNH-6270, is an angiotensin II receptor (AT1R) antagonist that is utilized in the treatment of high blood pressure. It is intended for research use only.
  • It is an angiotensin II receptor (AT1R) antagonist.
  • It is used to treat high blood pressure.
  • It inhibits the growth of HeLa cells in a concentration- and time-dependent manner.
  • Repeated dosing decreases mean arterial blood pressure without significantly influencing body weight or food intake.
  • Treatment inhibits cardiac hypertrophy.
  • Treatment reverses decreased gene expressions of ACE2 and Mas receptor, and inhibits enhanced NADPH oxidase (Nox)4 expression and reactive oxygen species.